Order Entry
Canada
ContactUsLinkComponent
Cyclo [-RGDyK(HiLyte™ Fluor 750)]
Cyclo [-RGDyK(HiLyte™ Fluor 750)]
Catalog # 103007-284
Supplier:  Anaspec Inc
CAS Number:  
Cyclo [-RGDyK(HiLyte™ Fluor 750)]
Catalog # 103007-284
Supplier:  Anaspec Inc
Supplier Number:  AS-62333-01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    0.1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    RGD conjugated Peptide|RGDyK Peptide|RGD Peptide|RGD-HiLyte Peptide
  • Protein/peptide name:
    Cyclo [-RGDyK(HiLyte™ Fluor 750)]
  • Purity:
    95%
  • Molecular weight:
    1630.5 Da
  • Sequence:
    Cyclo[-RGDy-K(Hilyte Fluor™ 750)]
  • Cat. no.:
    103007-284
  • Supplier No.:
    AS-62333-01

Specifications

About this item

Cyclo(-RGDy-K) is labeled with HiLyte™ Fluor 750 (Abs/Em=754/778 nm). This cyclic RGD peptide contains a subsituted d-Tyr instead of d-Phe on the fourth position. Substitution with Tyr results in a high affinity and selectivity for the avb3 integrin. Substitution with Tyr also allows for electrophilic radiohalogenation if desired.
Sequence:Cyclo[-RGDy-K(Hilyte Fluor™ 750)]
MW:1630.5 Da
% peak area by HPLC:95
Storage condition:-20° C