Order Entry
Canada
ContactUsLinkComponent
NF-kB Inhibitor
NF-kB Inhibitor
Catalog # 103008-418
Supplier:  Anaspec Inc
CAS Number:  
NF-kB Inhibitor
Catalog # 103008-418
Supplier:  Anaspec Inc
Supplier Number:  AS-64660
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    NF-κB cell-permeable inhibitory Peptide|SN50 Peptide
  • Protein/peptide name:
    NF-kB Inhibitor
  • Purity:
    95%
  • Molecular weight:
    2781.5 Da
  • Sequence:
    AAVALLPAVLLALLAPVQRKRQKLMP
  • Cat. no.:
    103008-418
  • Supplier No.:
    AS-64660

Specifications

About this item

SN50 is a NF-κB cell-permeable inhibitory peptide. it consists of the hydrophobic region of the signal peptide of Kaposi fibroblast growth factor (K-FGF) attached to a NF-kB p50 nuclear localization sequence (NLS). SN50 inhibits translocation of the NF-kB active complex into the nucleus.
Sequence:AAVALLPAVLLALLAPVQRKRQKLMP
MW:2781.5 Da
% peak area by HPLC:95
Storage condition:-20° C