Order Entry
Canada
ContactUsLinkComponent
Human MUC5AC-13
Human MUC5AC-13
Catalog # 103006-582
Supplier:  Anaspec Inc
CAS Number:  
Human MUC5AC-13
Catalog # 103006-582
Supplier:  Anaspec Inc
Supplier Number:  AS-61333
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    MUC5AC Peptide|human mucin MUC5AC Peptide
  • Protein/peptide name:
    MUC5AC-13
  • Purity:
    95%
  • Molecular weight:
    1704.6 Da
  • Sequence:
    GTTPSPVPTTST-T*-SAP (* = GalNAc-modified residue)
  • Cat. no.:
    103006-582
  • Supplier No.:
    AS-61333

Specifications

About this item

This glycopeptide is an N-acetyl galactosamine (GalNAc)-modified MUC5AC mucin peptide containing the single site of threonine 13 labeled with GalNAc (T*). Polypeptide N-acetylgalactosaminyltransferase (ppGaNTase) catalyzes the transfer of GalNAc from the nucleotide sugar UDP-GalNAc to threonine. The MUC5AC gene is mainly expressed in gastric and tracheo-bronchial mucosae, and some tumors.
Sequence:GTTPSPVPTTST-T*-SAP (* = GalNAc-modified residue)
MW:1704.6 Da
% peak area by HPLC:95
Storage condition:-20° C