Order Entry
Canada
ContactUsLinkComponent
 
Melan-A/MART-1 (27-35)
Catalog # 103003-258
Supplier:  Anaspec Inc
CAS Number:  
Melan-A/MART-1 (27-35)
Catalog # 103003-258
Supplier:  Anaspec Inc
Supplier Number:  AS-60608
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    MART Peptide|Melan-A Peptide|Melanocyte differentiation antigen Peptide
  • Protein/peptide name:
    Melan-A/MART-1 (27-35)
  • Purity:
    95%
  • Molecular weight:
    814.6 Da
  • Sequence:
    AAGIGILTV
  • Cat. no.:
    103003-258
  • Supplier No.:
    AS-60608

Specifications

About this item

Melan-A is a melanocyte differentiation antigen recognized by cytotoxic T lymphocytes. This sequence is a naturally presented Melan-A/MART-1 nonamer peptide. The Melan-A/MART-1 gene is expressed by normal melanocytes, as well as by most fresh melanoma samples and melanoma cell lines. This peptide appears to be a very common immunogenic epitope for HLA-A2-restricted melanoma-specific tumor-infiltrating lymphocytes (TIL) and may be useful for the development of immunotherapeutic strategies.
Sequence:AAGIGILTV
MW:814.6 Da
% peak area by HPLC:95
Storage condition:-20° C