Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:HumanRat
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:[Leu31|NPY|Pro34]-NPY (non-Y2 receptor agonist)
- Protein/peptide name:[Leu31, Pro34]-Neuropeptide Y
- Purity:95%
- Molecular weight:4240.8 Da
- Sequence:YPSKPDNPGEDAPAEDMARYYSALRHYINLLTRPRY-NH2
- Cat. no.:102996-554
- Supplier No.:AS-24366
Like neuropeptide Y (NPY) and other peptides of the family, this peptide adopts a folded hairpin structure with the terminal segments in close proximity. An intracerebroventricular injection of NPY or [Leu31, Pro34]-NPY (non-Y2 receptor agonist) given during middle cerebral artery occlusion increases the infarct volume and nitric oxide (NO) overproduction in the rat brain.
Sequence:YPSKPDNPGEDAPAEDMARYYSALRHYINLLTRPRY-NH2
MW:4240.8 Da
% peak area by HPLC:95
Storage condition:-20° C