Order Entry
Canada
ContactUsLinkComponent
Substance P, FAM (Carboxyfluorescein)
Substance P, FAM (Carboxyfluorescein)
Catalog # 103006-404
Supplier:  Anaspec Inc
CAS Number:  
Substance P, FAM (Carboxyfluorescein)
Catalog # 103006-404
Supplier:  Anaspec Inc
Supplier Number:  AS-61201
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    FAM (Carboxyfluorescein)
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Substance P neuropeptide FAM
  • Protein/peptide name:
    Substance P
  • Purity:
    95%
  • Molecular weight:
    1706 Da
  • Sequence:
    FAM-RPKPQQFFGLM-NH2
  • Cat. no.:
    103006-404
  • Supplier No.:
    AS-61201

Specifications

About this item

Substance P (SP) is an important neuropeptide belonging to the tachykinin family. It acts as a neurotransmitter and neuromodulator. It is closely related to neurokinin A, both originating from the same precursor preprotachykinin A. It is released from the terminals of sensory nerves and is involved in inflammation and pain processes. This peptide is a fluorescent (FAM)-labeled Substance P, Abs/Em = 494/521 nm.
Sequence:FAM-RPKPQQFFGLM-NH2
MW:1706 Da
% peak area by HPLC:95
Storage condition:-20° C