To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:Gastrin releasing Peptide receptor (GRPR) detecting Peptide|Bombesin Peptide
- Protein/peptide name:[Lys3]-Bombesin
- Purity:95%
- Molecular weight:1591.9 Da
- Sequence:Pyr-QKLGNQWAVGHLM-NH2
- Cat. no.:102999-360
- Supplier No.:AS-22838
PET (Positron Emission Tomography) imaging of [Lys3]-bombesin is able to detect gastrin-releasing peptide receptor (GRPR) positive prostate cancer. An immunoconjugate of [Lys3]-bombesin and corresponding monoclonal antibody can specifically induce (CD64)-dependent monocyte and neutrophil-mediated lysis of small cell carcinoma.
Sequence:Pyr-QKLGNQWAVGHLM-NH2
MW:1591.9 Da
% peak area by HPLC:95
Storage condition:-20° C