Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Rat
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:hypothalamic Peptide|Rfa Peptide|Rfa neuropeptide|Rfamide Peptide
- Protein/peptide name:26Rfa, Hypothalamic Peptide
- Purity:95%
- Molecular weight:2820.2 Da
- Sequence:ASGPLGTLAEELSSYSRRKGGFSFRF-NH2
- Cat. no.:103006-890
- Supplier No.:AS-61650
26Rfa is a neuropeptide belonging to the RFamide peptide family. It exhibits orexigenic activity and has been associated to obesity. The primary structures of human, rat, and frog 26RFa exhibit 80% identity, and the C-terminal octapeptide is fully conserved from amphibians to mammals.
Sequence:ASGPLGTLAEELSSYSRRKGGFSFRF-NH2
MW:2820.2 Da
% peak area by HPLC:95
Storage condition:-20° C