Order Entry
Canada
ContactUsLinkComponent
Mouse Myosin H Chain Fragment
Mouse Myosin H Chain Fragment
Catalog # 103007-394
Supplier:  Anaspec Inc
CAS Number:  
Mouse Myosin H Chain Fragment
Catalog # 103007-394
Supplier:  Anaspec Inc
Supplier Number:  AS-62554
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Myosin H Peptide|alpha-myosin chain Peptide|Myosin H Chain Peptide
  • Protein/peptide name:
    Myosin H Chain Fragment
  • Purity:
    95%
  • Molecular weight:
    2073.4 Da
  • Sequence:
    Ac-RSLKLMATLFSTYASADR
  • Cat. no.:
    103007-394
  • Supplier No.:
    AS-62554

Specifications

About this item

This peptide is a fragment of the murine heart muscle specific peptide derived from a-myosin H chain. This peptide was used for induction of autoimmune myocarditis.
Sequence:Ac-RSLKLMATLFSTYASADR
MW:2073.4 Da
% peak area by HPLC:95
Storage condition:-20° C