Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Biotin
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Dynorphin A Peptide|Dynorphin 1-17 biotin
- Protein/peptide name:Dynorphin A (1-17)
- Purity:95%
- Molecular weight:2373.8 Da
- Sequence:Biotin-YGGFLRRIRPKLKWDNQ
- Cat. no.:102999-774
- Supplier No.:AS-23978
Specifications
About this item
Dynorphins are a class of opioid peptides that arise from the precursor protein prodynorphin. Upon cleavage by proprotein convertase 2 (PC2), multiple active peptides are released: dynorphin A, dynorphin B, and α/β-neo-endorphin. Dynorphins exert their effects primarily through the κ-opioid receptor (KOR), a G-protein-coupled receptor. Dynorphin has been shown to be a modulator of pain response, involved in drug addiction and appetite control.
Sequence:Biotin-YGGFLRRIRPKLKWDNQ
MW:2373.8 Da
% peak area by HPLC:95
Storage condition:-20° C