If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:DiATOME manipulator for use with Diamond Knives
- Cat. no.:103307-056
- Supplier No.:DZ100-01
Specifications
About this item
Easy handling and mounting of section ribbons.
- Frozen hydrated biological samples (CEMOVIS)
- Room temperature sectioning of water sensitive samples
- Dry resin sectioning of biological samples for chemical analysis
- Dry sectioning of industrial samples such as polymers
The left manipulator moves a conductive fibre on which the sections are attached by electrostatic force; the growing ribbon is guided.
The right manipulator guides the grid very precisely below the section ribbon, the sections are attached to the carbon film with electrostatic force.
The manip is compatible with Leica cryochambers FC6 and FC7 equipped with a Crion ionizer/charger. Adaption on other cryochambers upon request.