Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:1-Step™ Chloronaphthol
- Size:250 mL
- Cat. no.:CAPI34012
- Application:Immunoblotting
- Supplier No.:34012
Specifications
About this item
The 1-Step™ chloronaphthol solution of 4-chloro-1-naphthol for the chromogenic detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.
- Pre-filtered and ready-to-use
- Storage at 4 °C
Chloronaphthol reacts with HRP to form a distinct blue to blue-purple product to localize and visualize the activity of peroxidase-conjugated antibodies on blots or fixed tissue samples. Its alcohol-solubility is a property that can be utilized in certain double-staining applications.