Order Entry
Canada
ContactUsLinkComponent
Pierce™ AminoLink™ Plus Micro Immobilization Kit, Thermo Scientific
Pierce™ AminoLink™ Plus Micro Immobilization Kit, Thermo Scientific
Catalog # CAPI20475
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ AminoLink™ Plus Micro Immobilization Kit, Thermo Scientific
Catalog # CAPI20475
Supplier:  Thermo Scientific
Supplier Number:  20475
CAS Number:  

Specifications

  • Description:
    AminoLink™ Plus Micro Immobilization Kit
  • Cat. no.:
    CAPI20475
  • Supplier No.:
    20475
  • Sufficient For:
    10 columns

Specifications

About this item

Pierce™ AminoLink™ Plus Micro Immobilization Kit enables covalent attachment of amine-containing ligands for use in antibody purification and other affinity protocols.

  • Ideal for coupling small amounts (25 to 250 µg) of purified antibodies, proteins and other amine-containing ligands
  • Covalent attachment of the protein to resin results in a leach-resistant affinity support
  • Fast and easy to use – perform wash and elution steps using a microcentrifuge
  • Prepared peptide resin can be used several times with no significant loss of capacity
  • Immobilization efficiency greater than 80%
Delivery: Kit contains AminoLink™ Plus Spin Columns (0.1 mL, 10 columns), Wash Solution (25 mL), Sodium Cyanoborohydride Solution 5M (0.5 mL), Quenching Buffer (60 mL), Elution Buffer (50 mL), PBS Pack (makes 500 mL) and Collection Tubes (200 tubes).