Order Entry
Canada
ContactUsLinkComponent
Rat Secretin
Rat Secretin
Catalogue # 103003-340
Fournisseur:  Anaspec Inc
Numéro CAS:  
Rat Secretin
Catalogue # 103003-340
Fournisseur:  Anaspec Inc
Supplier Number:  AS-60677
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Conjugation:
    Unconjugated
  • Species:
    Rat
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Secretin Peptide
  • Protein/peptide name:
    Secretin
  • Purity:
    95%
  • Molecular weight:
    3028.7 Da
  • Sequence:
    HSDGTFTSELSRLQDSARLQRLLQGLV-NH2
  • Cat. no.:
    103003-340
  • Supplier No.:
    AS-60677

Spécifications

À propos de cet article

Secretin, a 27-amino acid gastrointestinal peptide hormone, has been implicated in numerous regulatory events involving the pancreas, biliary tree, stomach, nerves and kidney. It is relatively conserved through evolution. Rat secretin differs from its porcine counterpart by a single glutamine-for-arginine substitution at position 14.
Sequence: HSDGTFTSELSRLQDSARLQRLLQGLV-NH2
MW: 3028.7 Da
% Peak area by HPLC: 95
Storage condition: -20° C