Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Biotin
- Species:Human
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Biotinylated ghrelin Peptide
- Protein/peptide name:Biotin-Ghrelin
- Purity:95%
- Molecular weight:3597.2 Da
- Sequence:Biotin-GS-S(n-octanoyl)-FLSPEHQRVQQRKESKKPPAKLQPR
- Cat. no.:103005-290
- Supplier No.:AS-29391
This Ghrelin peptide is biotinylated at the peptide N-terminus. Ghrelin is an appetite stimulating peptide hormone secreted by stomach P/D1-type cells in humans and circulates in the bloodstream during fasting conditions. Enzymatically n-octanoylated Serine3 of this 28-amino acid Ghrelin is the endogenous ligand specific for growth hormone secretagogue receptor 1a (GHSR1a). The GHSR1a receptor appears to be dedicated to Ghrelin and is widely expressed in different tissues.
Sequence: Biotin-GS-S(n-octanoyl)-FLSPEHQRVQQRKESKKPPAKLQPR
MW: 3597.2 Da
% Peak area by HPLC: 95
Storage condition: -20° C