Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Size:0.5 mg
- Storage conditions:–20 °C
- Protein synonyms:biotin GLP 1 (7-36)
- Protein/peptide name:Biotin-Glucagon-like Peptide 1, GLP-1 (7-36), amide
- Purity:95%
- Molecular weight:3524 Da
- Sequence:Biotin-HAEGTFTSDVSSYLEGQAAKEFIAWLVKGR-NH2
- Cat. no.:102996-380
- Supplier No.:AS-23586
This GLP-1 (7-36) amide, is labeled with biotin at the peptide N-terminus. GLP-1 (7-36) amide is an incretin hormone that causes glucose dependent release of insulin by pancreatic beta cells. It is the cleavage product of GLP-1 (1-36) amide peptide (Cat# AS-22460). Both GLP-1 (7-36) and GLP-1 (7-37)-Cat# AS-20761, also play roles in gastric motility (gastric emptying), on the suppression of plasma glucagon levels (glucose production) and possibly on the promotion of satiety and stimulation of glucose disposal in peripheral tissues independent of the actions of insulin. GLP-1 (7-36) has a short half life of less than 2 minutes, and like GIP, is rapidly degraded by the enzyme dipeptidyl peptidase IV (DPP-4), which is widely expressed in a number of sites, including the endothelial cells of small gut arterioles. DPP-4 degrades GLP-1 (7-36) into the non insulinotropic GLP-1 (9-36)-Cat# AS-65070 (some studies suggest it may have weak insulinotropic activity). As a result, the majority of GLP-1 (and GIP) is inactivated as an insulinotrope before reaching the systemic circulation.
Sequence: Biotin-HAEGTFTSDVSSYLEGQAAKEFIAWLVKGR-NH2
MW: 3524 Da
% Peak area by HPLC: 95
Storage condition: -20° C