Order Entry
Canada
ContactUsLinkComponent
Human IGRP Catalytic Subunit-related Protein (206-214)
Human IGRP Catalytic Subunit-related Protein (206-214)
Catalog # 103008-082
Supplier:  Anaspec Inc
CAS Number:  
Human IGRP Catalytic Subunit-related Protein (206-214)
Catalog # 103008-082
Supplier:  Anaspec Inc
Supplier Number:  AS-64431
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    IGCRP Peptide|Islet-specific Glucose-6-phosphatase Catalytic subunit Related Protein|Islet specific catalytic subunit Peptide|islet-specific glucose-6-phosphatase catalytic subunit related protein 206-214|Islet catalytic subunit Peptide|IGRP 206-214
  • Protein/peptide name:
    IGRP Catalytic Subunit-related Protein (206-214)
  • Purity:
    95%
  • Molecular weight:
    1096.3 Da
  • Sequence:
    VYLKTNVFL
  • Cat. no.:
    103008-082
  • Supplier No.:
    AS-64431

Specifications

About this item

This peptide corresponds to residues 206–214 of murine islet-specific glucose-6-phosphatase catalytic subunit–related protein (IGRP). This peptide is T cells specific for proinsulin and IGRP induces diabetes in non-obese diabetic (NOD) mice.
Sequence: VYLKTNVFL
MW: 1096.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C