Order Entry
Canada
ContactUsLinkComponent
40Gap 27, Connexin Mimetic
40Gap 27, Connexin Mimetic
Catalog # 103007-448
Supplier:  Anaspec Inc
CAS Number:  
40Gap 27, Connexin Mimetic
Catalog # 103007-448
Supplier:  Anaspec Inc
Supplier Number:  AS-62641
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    GAP27|Connexin|GAP27 mimetic
  • Protein/peptide name:
    40Gap 27, Connexin Mimetic
  • Purity:
    95%
  • Molecular weight:
    1289.5 Da
  • Sequence:
    SRPTEKNVFIV
  • Cat. no.:
    103007-448
  • Supplier No.:
    AS-62641

Specifications

About this item

This peptide corresponds to the GAP27 domain of the second extracellular loop of dominant vascular connexin (Cx40), designated as 40Gap 27. It was used to investigate mechanisms through which oxidant stress impairs communication via gap junctions. When administered, 40Gap27 attenuates endothelium-dependent subintimal smooth muscle hyperpolarization.
Sequence:SRPTEKNVFIV
MW:1289.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C