Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Platelet C18G|Platelet factor|Platelet factor C18G|Platelet factor IV
- Protein/peptide name:Platelet Factor IV 18
- Purity:95%
- Molecular weight:2043.7 Da
- Sequence:ALYKKLLKKLLKSAKKLG
- Cat. no.:103007-328
- Supplier No.:AS-62412
C18G is a synthetic α-helical peptide derived from human platelet factor IV. This peptide was found to be antibacterial and is active against Salmonella.
Sequence:ALYKKLLKKLLKSAKKLG
MW:2043.7 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C