Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:CSVTCG thrombospondin|Thrombospondin Peptide|Cyclic Thrombospondin|Cyclic TSP
- Protein/peptide name:Thrombospondin-derived Peptide
- Purity:95%
- Molecular weight:566.7 Da
- Sequence:CSVTCG (S-S Bonded)
- Cat. no.:103009-490
- Supplier No.:AS-65371
This peptide is derived from thrombospondin and represents a binding motif responsible for thrombospondin-CD36 interaction. It is cyclized through a disulfide bond. Thrombospondin is a matrix-bound glycoprotein involved in cancer metastasis, tumor adhesion, and angiogenesis. This peptide has been shown to competitively inhibit platelet aggregation and tumor metastasis.
Sequence:CSVTCG (S-S Bonded)
MW:566.7 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C