Order Entry
Canada
ContactUsLinkComponent
Bovine Neurotensin Peptide
Bovine Neurotensin Peptide
Catalog # 103007-476
Supplier:  Anaspec Inc
CAS Number:  
Bovine Neurotensin Peptide
Catalog # 103007-476
Supplier:  Anaspec Inc
Supplier Number:  AS-62677
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Bovine
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Neurotensin bioactive Peptide
  • Protein/peptide name:
    Neurotensin Peptide
  • Purity:
    95%
  • Molecular weight:
    1672.92 Da
  • Sequence:
    pE-LYENKPRRPYIL
  • Cat. no.:
    103007-476
  • Supplier No.:
    AS-62677

Specifications

About this item

This 13 amino acid peptide was first isolated from bovine hypothalamus and was named from its neuronal localization. It was detected in the central nervous system (CNS) and peripheral tissues mainly in the gastrointestinal tract. This bioactive form of neurotensin post-translationally modified at a Glu residue was isolated from porcine intestine.
Sequence:pE-LYENKPRRPYIL
MW:1672.92 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C