Order Entry
Canada
ContactUsLinkComponent
Mouse BNP-45, AnaSpec
Mouse BNP-45, AnaSpec
Catalog # 102971-866
Supplier:  Anaspec
CAS Number:  
Mouse BNP-45, AnaSpec
Catalog # 102971-866
Supplier:  Anaspec
Supplier Number:  AS-61152
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Mouse
  • Size:
    0.5 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Brain Natriuretic Peptide-45
  • Protein/peptide name:
    BNP-45
  • Purity:
    95%
  • Molecular weight:
    4919.6 Da
  • Sequence:
    SQGSTLRVQQRPQNSKVTHISSCFGHKIDRIGSVSRLGCNALKLL (Disulfide bridge:23 - 39)
  • Cat. no.:
    102971-866
  • Supplier No.:
    AS-61152

Specifications

About this item

BNP-45 represents the 45 amino acids at the C-terminus and the mouse BNP-45 has all the amino acid residues thought essential for NP bioactivity, although sequence identity when studied with other BNP hormones (rat, 64%; dog, 53%; pig, 50%; and human, 44%) was clearly less than the identity among ANF hormones. The threonine 81 residue at which a protein kinase C phosphorylation site is present in the putative mature mouse BNP-45 hormone is not conserved in the rat sequence.
Sequence:SQGSTLRVQQRPQNSKVTHISSCFGHKIDRIGSVSRLGCNALKLL (Disulfide bridge:23 - 39)
MW:4919.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C