Order Entry
Canada
ContactUsLinkComponent
Human Brain Natriuretic Peptide 32
Human Brain Natriuretic Peptide 32
Supplier:  Anaspec Inc
Certificates
Human Brain Natriuretic Peptide 32
Supplier:  Anaspec Inc

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    human|Brain Natriuretic Peptide-32
  • Protein/peptide name:
    Brain Natriuretic Peptide 32
  • Purity:
    95%
  • Molecular weight:
    3464.1 Da
  • Sequence:
    SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH (Disulfide bridge: 10-26)

Specifications

Related Information

Frequently Bought Together