Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:5 mg
- Storage conditions:–20 °C
- Protein synonyms:Kallidin|Bradykinin-Lys0
- Protein/peptide name:[Lys0]-Bradykinin (Kallidin)
- Purity:95%
- Molecular weight:1188.4 Da
- Sequence:KRPPGFSPFR
- Cat. no.:102999-364
- Supplier No.:AS-22853
[Lys0]-BK(1-10) is one of the two main Bradykinin peptides that act as a mediator of inflammation with evidence showing its release at high nanomolar concentrations into the tear-film of ocular allergic patients.
Sequence:KRPPGFSPFR
MW:1188.4 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C