Order Entry
Canada
ContactUsLinkComponent
Human Fibrinopeptide A
Human Fibrinopeptide A
Catalog # 102996-098
Supplier:  Anaspec Inc
CAS Number:  
Human Fibrinopeptide A
Catalog # 102996-098
Supplier:  Anaspec Inc
Supplier Number:  AS-20735
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    FPA
  • Protein/peptide name:
    Fibrinopeptide A
  • Purity:
    95%
  • Molecular weight:
    1536.6 Da
  • Sequence:
    ADSGEGDFLAEGGGVR
  • Cat. no.:
    102996-098
  • Supplier No.:
    AS-20735

Specifications

About this item

Fibrinopeptide A is a 16-amino acid cleavage product of thrombin-induced proteolytic cleavage of fibrinogen. Liberation of FPA and another 14-amino acid peptide, fibrinopeptide B, uncovers the E domain of fibrinogen. The residual protein, fibrin monomer, polymerizes to form fibrin clot. Thus, liberation of approximately 4 ng/ml of FPA per milligram of fibrinogen is closely linked to clot formation. Elevation of Fibrinopeptide A levels in plasma is seen in association with disorders such as disseminated intravascular coagulation, deep venous thrombosis, arterial thrombosis, and malignancy.
Sequence:ADSGEGDFLAEGGGVR
MW:1536.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C