Order Entry
Canada
ContactUsLinkComponent
Pierce™ Mitochondria Isolation Kit for Cultured Cells, Thermo Scientific
Pierce™ Mitochondria Isolation Kit for Cultured Cells, Thermo Scientific
Catalog # CAPI89874
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Mitochondria Isolation Kit for Cultured Cells, Thermo Scientific
Catalog # CAPI89874
Supplier:  Thermo Scientific
Supplier Number:  89874
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mitochondria Isolation Kit for Cultured Cells
  • Cat. no.:
    CAPI89874
  • Application:
    Intact Mitochondria from Cultured Mammalian Cells
  • Supplier No.:
    89874
  • Sufficient For:
    50 Samples each containing 20 Million Cultured Mammalian Cells

Specifications

About this item

The Mitochondria Isolation Kit for Cultured Cells provides a versatile, microcentrifuge-tube method for fractionating intact mitochondria from cultured mammalian cell samples.

  • Fast- isolate intact mitochondria from 20 million cells in approximately 40 minutes (post-cell harvest)
  • Flexible - process several samples simultaneously using the reagent-based method, or obtain highest possible yield with the Dounce homogenization method
  • Optimizable - guidelines provided for optimizing purity vs. yield with the reagent-based method
  • Benchtop-easy - isolation performed in a microcentrifuge tube
  • Compatible - isolated mitochondria may be lysed with detergent or processed for almost any downstream application, including 2D electrophoresis

The Mitochondria Isolation Kit uses a non-mechanical, reagent-based method that allows multiple (six) cultured cell samples to be processed concurrently in about 40 minutes

The product instructions describe guidelines for optimizing purity vs. yield parameters. The kit also offers an optimized Dounce homogenization procedure, which results in two-fold greater mitochondria recovery compared to the reagent-based method. Both methods use differential centrifugation to separate the mitochondrial and cytosolic fractions with a benchtop microcentrifuge and are completed in approximately 40 minutes (post-cell harvest).

Once isolated, the mitochondria can be used in downstream applications such as apoptosis, signal transduction and metabolic studies, as well as to facilitate mitochondrial proteomics efforts.

Cultured mammalian cell pellets are gently lysed using a proprietary formulation that results in maximum yield of mitochondria with minimal damage to integrity.

Delivery: Kit contains Mitochondria isolation reagent A (50 mL), Mitochondria isolation reagent B (0.5 mL) and Mitochondria isolation reagent C (65 mL).