Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:Melanoma gp 25-33|Melanoma antigen gp100 (25–33)|Melanoma gp100
- Protein/peptide name:gp100 (25–33) Peptide
- Purity:95%
- Molecular weight:1155.3 Da
- Sequence:KVPRNQDWL
- Cat. no.:103007-408
- Supplier No.:AS-62589
This is amino acids 25 to 33 fragment of human melanoma antigen gp100. This H-2Db restricted epitope is recognized by T cells. The gp100-specific, H-2Db-restricted, CD8+ T cells are capable of recognizing B16 melanoma but not normal melanocytes. This peptide was used as an immunogen in multiple cancer immunotherapy studies.
Sequence: KVPRNQDWL
MW: 1155.3 Da
% Peak area by HPLC: 95
Storage condition: -20°C