Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:HIV
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:NSF22sscr Peptide|(TAT) N-ethylmaleimide-sensitive factor (NSF)|TAT NSF Peptide|TAT NSFs22scr
- Protein/peptide name:TAT-NSF222scr Fusion Polypeptide
- Purity:95%
- Molecular weight:4214.9 Da
- Sequence:YGRKKRRQRRR-GGG-ENSFRFLADIFPAKAFPVRFE
- Cat. no.:103007-246
- Supplier No.:AS-62210
This is a scrambled TAT-NSF222scr fusion polypeptide. It is composed of 11 amino acids from the cell permeable human immunodeficiency virus TAT polypeptide, 3 glycines as a linker, followed by scrambled N-Ethyl-maleimide-sensitive factor (NSF) D1 domain. This peptide is used as a control for the TAT-NSF222 peptide.
Sequence: YGRKKRRQRRR-GGG-ENSFRFLADIFPAKAFPVRFE
MW: 4214.9 Da
% Peak area by HPLC: 95
Storage condition: -20°C