Order Entry
Canada
ContactUsLinkComponent
HIV Tat-NR2Bct Peptide
HIV Tat-NR2Bct Peptide
Catalog # 103007-148
Supplier:  Anaspec Inc
CAS Number:  
HIV Tat-NR2Bct Peptide
Catalog # 103007-148
Supplier:  Anaspec Inc
Supplier Number:  AS-62042
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    HIV
  • Size:
    1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Tat decoy|post synaptic Peptide|Tat-NR2Bct|NR2Bct|PSD decoy Peptide
  • Protein/peptide name:
    Tat-NR2Bct Peptide
  • Purity:
    95%
  • Molecular weight:
    2518.9 Da
  • Sequence:
    YGRKKRRQRRRKLSSIESDV
  • Cat. no.:
    103007-148
  • Supplier No.:
    AS-62042

Specifications

About this item

This is the membrane-permeable postsynaptic density (PSD)-95-binding (decoy) peptide Tat-NR2Bct. It can transduce into neurons in cell culture.
Sequence: YGRKKRRQRRRKLSSIESDV
MW: 2518.9 Da
% Peak area by HPLC: 95
Storage condition: -20°C