Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Chicken
- Size:1 mg
- Storage conditions:–20 °C
- Protein synonyms:OVA Peptide|OVA 329-337 Peptide
- Protein/peptide name:OVA (329-337)
- Purity:95%
- Molecular weight:925 Da
- Sequence:AAHAEINEA
- Cat. no.:103008-446
- Supplier No.:AS-64777
Specifications
About this item
This sequence is OVA (ovalbumin) peptide residues 257 to 280, also known as OT-II (OVA-specific T-cell receptor) peptide, the core H2b-restricted class II MHC epitope.
Sequence: AAHAEINEA
MW: 925 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C