Order Entry
Canada
Orders LinkContactUsLinkComponent
R-UNIVERSAL Epitope Recovery Buffer, Electron Microscopy Sciences

R-UNIVERSAL Epitope Recovery Buffer, Electron Microscopy Sciences

Recovery buffer (10X stock), 125 ml
Catalog # 103305-558
Supplier #:  62719-10
R-UNIVERSAL Epitope Recovery Buffer, Electron Microscopy Sciences
Catalog # 103305-558
CAS Number:  
R-UNIVERSAL Epitope Recovery Buffer, Electron Microscopy Sciences
R-UNIVERSAL Epitope Recovery Buffer, Electron Microscopy Sciences

Recovery buffer (10X stock), 125 ml

CAS Number:  
Price
$505.64
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Epitope recovery on formalin-fixed, paraffin embedded tissue sections, requires heat-induced treatment in buffer or sometimes, proteolytic treatment of the deparaffinized tissue section.

  • Clear, non-toxic
  • Store at 4 °C

EMS offers a novel (patent-pending) technology for epitope recovery, primarily based on reversing the fixation effects of formaldehyde, which created links primarily between ε-amino groups of lysine and other amino-groups.

For epitope recovery dilute 1 part of stock with 9 parts of deionized water.

Note: Stable for 1 year stored unopened and after opening can store up to 6 months.

Certifications: DIN EN ISO 9001:2008.

Specifications

Specifications

  • Description:
    Recovery buffer (10X stock)
  • Size:
    125 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.