Rapid-Core Cutting Mat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Composite cutting mats for coring samples.
- Protects tips and working surface
- Reversible, inert and self-healing
- Sterilized with ethanol
Five layer composite cutting mat will cut cleanly through an object and into the mat. Ensures the cored sample comes out completely without burrs and bits.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together