Double FIB Sample and Grid Holder, Electron Microscopy Sciences
Product Details & Documents
Double FIB sample and grid holder holds sample mounted on two standard pin stubs for FIB milling and lift-out procedures.
- Vacuum grade aluminum with brass screws
- Conveniently holds two FIB grids close to the sample to mount prepared TEM lamellae on the FIB grid for TEM imaging
- Cost-effective and suitable for all FIB/SEM systems which accept pin mount holders, including the FEI, ZEISS and Tescan systems
Overall dimensions: 36.5 x 12.7 x 11.6 mm (1.44" x 0.5" x 0.46")
Pin: Standard 3.2 mm (1/8")
Delivery information: Includes Philips screwdriver.
Specifications
- Description:Double FIB Sample and Grid Holder, Pin
- Dimensions:1/2" (12.7 mm)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



