Orders LinkContactUsLinkComponent
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
EDTA Buffer (10X), pH 8.5, Electron Microscopy Sciences
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.
In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC
Preparation of Working Solution: Dilute this 10X buffer as needed (90 mL of deionized or distilled water+10 mL of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.
The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Product Family Options
  • Description
    Size
    pH
  • EDTA Buffer
    100 mL
    8.5
    Each (100ml)
    Each (100ml) - $191.72
    $191.72
    Each (100ml)
       $191.72
     : 64142-09
  • EDTA Buffer
    1000 mL
    8.5
    Each (1,000ml)
    Each (1,000ml) - $390.61
    $390.61
    Each (1,000ml)
       $390.61
     : 64142-10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.