If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:4" Holes Bottle Carrier
- Cat. no.:100498-722
- Supplier No.:64371-04
Benchtop wash bottle holder prevents spills.
- Protects bench top from spills and stains
- Easy access to solvent wash bottles
- Made from chemical resistant plastic that is easy to clean
For the storage or carriage of wash bottles or solvents. Six-pack storage, convenient carrying handles. Sturdy construction and they are resistant to solvents. 13.5×10×11” H.
Available in two sizes: 4" holes (white) and 27/8" holes.