About this item
Aß (1-28) is highly hydrophilic and shares sequences with bA4, the major component of Aß. Its assembly is fibrillar, i.e., elongated in a single direction. Reports show that synthetic peptides Aß (1-40) and Aß (1-28) have significant effects on normal human plasma cholesterol esterification rate.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNK
Molecular Weight: 3262.5 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage conditions:–20 °C
- Protein synonyms:amyloid 1-28|amyloid beta (1-28)|abeta (1-28)|beta amyloid 1-28
- Protein/peptide name:beta-Amyloid (1-28)
- Purity:95%
- Molecular weight:3262.5 Da
- Sequence:DAEFRHDSGYEVHHQKLVFFAEDVGSNK