Order Entry
Canada
ContactUsLinkComponent
Human Beta-Amyloid (1-40)-Lys(LC-biotin)-NH2, Biotin
Human Beta-Amyloid (1-40)-Lys(LC-biotin)-NH2, Biotin
Catalog # 103007-108
Supplier:  Anaspec Inc
CAS Number:  
Human Beta-Amyloid (1-40)-Lys(LC-biotin)-NH2, Biotin
Catalog # 103007-108
Supplier:  Anaspec Inc
Supplier Number:  AS-61962-01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Species:
    Human
  • Size:
    0.1 mg
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    amyloid 1-40 Lys(Biotin) LC FAM|beta amyloid (1-40) Lys(Biotin LC) FAM|amyloid beta (1-40) Lys(Biotin) LC FAM|abeta (1-40) Lys(Biotin) LC FAM
  • Protein/peptide name:
    beta-Amyloid (1-40)-Lys(LC-biotin)-NH2
  • Purity:
    95%
  • Molecular weight:
    5154.9 Da
  • Sequence:
    5-FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV-K(LC-BIOTIN)-NH2
  • Cat. no.:
    103007-108
  • Supplier No.:
    AS-61962-01

Specifications

About this item

This is a 5-FAM labeled ß-?Amyloid (1-40). This amidated peptide is also biotinylated on the lysine side chain with 6-aminohexanoate (LC) as a spacer, Ex/Em=490 nm/ 520 nm.
Sequence: 5-FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV-K(LC-BIOTIN)-NH2
Molecular Weight: 5154.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C