NOS Activity Assay Kit, Cayman Chemical Company
75817-234
:
- Brand name:CAYM
- Cat. no.:75817-234
- Contents:Kit contains: Positive Control, Buffer, Reagents, Spin Columns, and Collection Tubes
- Supplier No.:781001-1
For the measurement of NOS activity.
- Measure NOS activity in cell lysates and purified preparations
- Material for 50 reactions
- Radioactive substrate not supplied
The NOS Activity Assay measures NOS activity by monitoring the conversion of radiolabeled arginine to citrulline. This assay is simple, sensitive, and specific for NOS activity and can be used with both crude and purified enzyme preparations. The kit includes sufficient materials and reagents for 50 total reactions. Radiolabeled arginine and NADPH are not included with the kit.