Order Entry
ContactUsLinkComponent
COX (ovine/human) Inhibitor Screening Assay Kit, Cayman Chemical Company
COX (ovine/human) Inhibitor Screening Assay Kit, Cayman Chemical Company
Catalog #: 75816-930
CAS Number:  
KIT 96 WL COX(OVN/HU) INH SCRG 405-420NM
COX (ovine/human) Inhibitor Screening Assay Kit, Cayman Chemical Company
Catalog #: 75816-930
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Sensitivity:
    80% B/B0: 29 pg/ml
  • Brand name:
    Cayman
  • Cat. no.:
    75816-930
  • Contents:
    Kit contains: Antiserum, Tracer, Standard, Buffers, Polysorbate 20, Reagent, COX-2 (human recombinant), Heme, Arachidonic Acid (substrate), Potassium Hydroxide, Hydrochloric Acid, Stannous Chloride, and Plate
  • Range:
    15.6 - 2,000 pg/mL
  • Supplier No.:
    560131-96

Specifications

About this item

Complete drug discovery assay for screening COX-1 and COX-2 inhibitors

  • Screen for inhibitors of ovine COX-1 or human COX-2
  • Directly measure PGF2α produced in the COX reaction
  • Assay 15 samples for COX-1 or COX-2 in duplicate
  • Plate-based colorimetric measurement (405-420 nm)
  • Midpoint: 50% B/B0: 125-250 pg/ml

The COX (ovine/human) Inhibitor Screening Assay directly measures PGF2α produced by SnCl2 reduction of COX-derived PGH2. The prostanoid product is quantified via ELISA using a broadly specific antibody that binds to all the major PG compounds. Thus, the Cayman COX Assay is more accurate and reliable than an assay based on peroxidase inhibition. Cayman’s COX Inhibitor Screening Assay includes both ovine COX-1 and human recombinant COX-2 enzymes in order to screen isozyme-specific inhibitors. This assay is an excellent tool which can be used for general inhibitor screening, or to eliminate false positive leads generated by less specific methods.

Customers Who Bought This Also Bought