Order Entry
Canada
ContactUsLinkComponent
Resolvin D1 ELISA Kit, Cayman Chemical Company
Resolvin D1 ELISA Kit, Cayman Chemical Company
Certificates
undefined
Resolvin D1 ELISA Kit, Cayman Chemical Company

Specifications

  • Assay Type:
    Competitive
  • Detection range:
    3.3 – 2000 pg/ml
  • Sensitivity:
    80% B/B0: 15 pg/ml

Specifications

Related Information

Frequently Bought Together