Specifications
- Brand name:Cayman
- Cat. no.:75816-674
- Contents:Kit contains: Hydrochloric Acid, DNPH, Solution, Guanidine Hydrochloride, Ethanol, Ethyl Acetate, and Plate
- Supplier No.:10005020-96
Specifications
About this item
Convenient colorimetric assay for measurement of oxidized proteins
- Measure protein carbonyl content in plasma, serum, urine, tissue homogenates, and cell lysates
- Assay 24 samples in duplicate
- Plate-based colorimetric measurement (360-385 nm)
Cayman’s Protein Carbonyl Colorimetric Assay is a colorimetric assay for the measurement of oxidized proteins. The carbonyls of protein samples are derivatized using 2,4-dinitrophenylhydrazine. Formation of a Schiff base produces the corresponding hydrazone which can be analyzed spectrophotometrically at 360-385 nm. This assay can be used to measure oxidized protein in plasma, serum, cell lysates, and tissue homogenates.