Peg Racks for Test Tubes, Globe Scientific
Supplier: Globe Scientific
Jump to:
About this item
This versatile rack can be used as a tandard test tube rack or even be used as a drying rack for inverted tubes.
- Fully autoclavable
- Manufactured from a durable reinforced polypropylene
- Can be used to dry inverted tubes while minimizing collection of airborne contaminants
The peg design of the rack allows for easy viewing of the tube contents.
6 Options Available Below
Product Details & Documents
This versatile rack can be used as a tandard test tube rack or even be used as a drying rack for inverted tubes.
- Fully autoclavable
- Manufactured from a durable reinforced polypropylene
- Can be used to dry inverted tubes while minimizing collection of airborne contaminants
The peg design of the rack allows for easy viewing of the tube contents.
Documents
Product Family Options
13 mm
Product Information
- ColorNo. of HolesDimensionsAvailabilityPrice
- Supplier Number: 456150B
- Supplier Number: 456150R
- Supplier Number: 456150W
17 mm
Product Information
- ColorNo. of HolesDimensionsAvailabilityPrice
- Supplier Number: 456155B
- Supplier Number: 456155R
- Supplier Number: 456155W