Reagent Reservoirs, 12 Channel, DWK Life Sciences
Catalog #: 75800-888
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:48 ml
- Description:Reagent Reservoirs, 12 Channel
- Material:Polystyrene
- Cat. no.:75800-888
- Supplier No.:W845902
Specifications
About this item
Non sterile reagent reservoir made from PS.
Customers Who Bought This Also Bought