Autoclave Bag Clip, Chemglass
Catalog # 30622-744
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Autoclave Bag Clip
- Cat. no.:30622-744
- Supplier No.:CLS-1192-C001
Specifications
About this item
These open and close autoclave bag clip holds the processed bag closed for final disposal.
- Closed internal dimension of the clip is 7/16"
- Clip permits bag to safely vent internal pressure during autoclaving
- Holds filled autoclaved bags closed during cycle