Orders LinkContactUsLinkComponent
Drug Testing Model Set
Drug Testing Model Set
Catalog # 470232-070
Supplier:   Mega Molecules
 :  FC125
Drug Testing Model Set
Catalog # 470232-070
Supplier:  Mega Molecules
Drug Testing Model Set
Drug Testing Model Set
Drug Testing Model Set
Price
$39.45
Log in for availability
Each
Product Details & Documents
Hands on representation of drug testing models.
  • Includes 1 activity
  • Perfect for high school students
In this activity, students act as members of a hospital administration who are responsible for developing an effective substance-abuse prevention and drug testing program. Students will explore the chemical and physical properties of five illicit drugs and build a model of each drug. Students will analyze the results of urine test data and evaluate drug use among hospital applicants and employees.
Specifications
  • Description:
    Drug Testing Model Set