Orders LinkContactUsLinkComponent
Storage Vials, Quick/Snap, MicroSolv
Storage Vials, Quick/Snap, MicroSolv
Storage Vials, Quick/Snap, MicroSolv
Certificates
Storage Vials, Quick/Snap, MicroSolv
Storage Vials, Quick/Snap, MicroSolv
4 Options Available Below
Product Details & Documents
These easy to use storage vials and caps use a push on technology for ease and comfort.
  • Easy to handle
  • Quick and easy
  • No liners required in the cap
Use these vials to store samples that are not critical.
Ordering information: Caps sold separately.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Cap included
    Cap Size
    Availability
    Price
  • 97058-138
    Vials, quick snap storage
    Vials, quick snap storage
    5.00 ml
    40×22 mm
    No
    ND18
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $63.12
    $63.12
    Pack of 100
       $63.12
    Supplier Number: 97010-CV05
  • 97058-132
    Vials, quick snap storage
    Vials, quick snap storage
    10.00 ml
    50×22 mm
    No
    ND18
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $83.18
    $83.18
    Pack of 100
       $83.18
    Supplier Number: 97010-CV10
  • 97058-134
    Vials, quick snap storage
    Vials, quick snap storage
    15.00 ml
    48×26 mm
    No
    ND22
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $112.98
    $112.98
    Pack of 100
       $112.98
    Supplier Number: 97010-CV15
  • 97058-136
    Vials, quick snap storage
    Vials, quick snap storage
    25.00 ml
    65×26 mm
    No
    ND22
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $115.53
    $115.53
    Pack of 100
       $115.53
    Supplier Number: 97010-CV25
Caps
Product Information
  • Catalog #
    Description
    Cap type
    Cap color
    Availability
    Price
  • Small caps, use with ND18 vials, 19.8 x 5.2 mm
    PE-push on cap
    Transparent
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $18.72
    $18.72
    Pack of 100
       $18.72
Product Information
  • Catalog #
    Description
    Cap type
    Cap color
    Availability
    Price
  • Large caps, use with ND22 vials, 23.5 x 5.5 mm
    PE-push on cap
    Transparent
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $19.56
    $19.56
    Pack of 100
       $19.56