ContactUsLinkComponent
Type 70 Ti Fixed-Angle Rotor, Titanium, Beckman Coulter®
Type 70 Ti Fixed-Angle Rotor, Titanium, Beckman Coulter®
Catalog # 101946-786
Supplier:  Beckman Coulter
Product available on GSA Advantage®
Type 70 Ti Fixed-Angle Rotor, Titanium, Beckman Coulter®
Catalog # 101946-786
Supplier:  Beckman Coulter
Supplier Number:  337922
Product available on GSA Advantage®
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Cat. no.:
    101946-786
  • Supplier No.:
    337922
Specifications
About this item
For use in instruments classified: H R S.
Applications: Differential centrifugation of subcellular particles.
Delivery: Type 70 Ti Rotor assembly with 3 each O-rings for rotor handle, O-rings for rotor lid, and 2 each overspeed disk, 70,000 rpm.