ContactUsLinkComponent
Staticmaster® Electrostatic Discharge Workstation Kit with I-Beam Ionizer, NRD
Staticmaster® Electrostatic Discharge Workstation Kit with I-Beam Ionizer, NRD
Catalog # 75794-896
Supplier:  NRD Staticmaster
undefined
Staticmaster® Electrostatic Discharge Workstation Kit with I-Beam Ionizer, NRD
Catalog # 75794-896
Supplier:  NRD Staticmaster
Supplier Number:  SMPF-KITELEC
Specifications
  • Description:
    ESD workstation kit with I-beam ionizer
  • Cat. no.:
    75794-896
  • Supplier No.:
    SMPF-KITELEC
Specifications
About this item
This Staticmaster® powder/filter AC kit uses the beam with power supply to provide electrical ionization, giving it the ability to remove static charge on any material introduced into the ion field generated by the I-Beam.
  • Provides electrical ionization
  • Removes static charge on any material introduced into ion field
Includes the ESD workstation kit.