Fat/Lipid Tests
Lipid Test Strips, 75
Catalog # 470093-682
Supplier: Precision Laboratories
Supplier #: LIP-1B-75
Jump to:
Product Details & Documents
A simple dip test for presence of lipids in solution.
- Resealable zip closure bag
- Waterproof chart
Easily test solutions for the presence of fats with these single use dip tests. Each strip had a single testing pad which will change color in the presence of lipids. Simply dip, wait 2 seconds and compare to the chart.
Specifications
- Description:Lipid Test Strips
- No. of tests:75