About this item
The sequence (Accession # NP_003076)) corresponds to the human β-synuclein along with a 6x His tag expressed in E. coli. This human β-synuclein recombinant protein is offered at a low endotoxin level of <0.1 EU per μg protein.
Beta-synuclein belongs to the family of highly conservative proteins in vertebrates. N-terminal of β-synuclein is highly homologous to α-, γ-synucleins and consists of degenerative “KTKEGV” repeats. Similar to α-synuclein, beta-synuclein is found primarily in the brain; however, it does not associate with Lewy bodies in Parkinson disease like α-synuclein. Beta-synuclein was found to inhibit production of phosphatidic acid by the phospholipase D2 transmembrane protein in vitro. In addition, beta-synuclein was detected in many breast and ovarian tumors. Recent investigations demonstrated that beta-synuclein can induce mild experimental autoimmune encephalomyelitis (EAE) in Lewis rats.
The human β-synuclein recombinant proteins from AnaSpec are offered with low endotoxin levels of <1 EU per μg protein.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 2-4 °C for immediate use within 1 week or at –80 °C for up to 6 months. Avoid repeated freeze-thaw cycles.
- Gene ID:NP_003076
- Protein synonyms:SNCB|Recombinant β-Synuclein|low endotoxin synuclein|Human SNCB|low endotoxin beta-Synuclein|β-Synuclein|Human β-Synuclein
- Protein/peptide name:beta-Synuclein
- Purity:> 90% as determined by SDS-PAGE